Autosampler Syringes with Removable Needle for Bruker (Varian) CP-8400/8410, CP-9010/9050
Autosampler Syringes with Removable Needle for Bruker (Varian) CP-8400/8410, CP-9010/9050
Actual product may vary
Autosampler Syringes with Removable Needle for Bruker (Varian) CP-8400/8410, CP-9010/9050
For reduced non specific sample interaction and increased analyte recovery
Have Questions?
Catalog NumberNeedle GaugeConnection TypeDiameter (English) Outer NeedleModel
SGE00295226 GRemovable0.018 in.10R-VA8400-5/0.47C
Catalog number SGE002952
Price (MYR)
422.00
EA
Add to cart
Needle Gauge:
26 G
Connection Type:
Removable
Diameter (English) Outer Needle:
0.018 in.
Model:
10R-VA8400-5/0.47C
Price (MYR)
422.00
EA
Add to cart
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Needle Gauge26 G
Diameter (English) Outer Needle0.018 in.
Diameter (Metric) Outer Needle0.47 mm
For Use With (Application)Gas chromatography
Length (English) Needle1.9 in.
Product TypeAutosampler Syringe
Model10R-VA8400-5/0.47C
QuantityEa.
Volume (Metric)10 μL
Length (Metric) Needle50 mm
Needle Point StyleCone
For Use With (Equipment)Bruker/Varian CP-8400/8410, CP-9010/9050
Connection TypeRemovable
Unit SizeEA