Check Order Status
Microvolume Syringes
Actual product may vary
Microvolume Syringes
Plunger protection minimizes risk of bending
Have Questions?
Catalog Number
Needle Gauge
Length (English)
Length (English) Needle
Needle Point Style
SGE002839
23 G
1.65 in.
1.65 in.
Cone
Catalog number
SGE002839
Price (MYR)
462.00
EA
Add to cart
Needle Gauge:
23 G
Length (English):
1.65 in.
Length (English) Needle:
1.65 in.
Needle Point Style:
Cone
Price (MYR)
462.00
EA
Add to cart
Extended borosilicate glass barrel with reinforced plunger for additional plunger protection
Accuracy and reproducibility of ±1%
Volume: 10μL
Barrel O.D.: 6.5mm, 8mm for guided plunger
Scale length: 54.1mm
For making microvolume injections of 1.5μL to 8μL
Specifications
Material
Borosilicate Glass
Needle Gauge
23 G
Diameter (Metric) Outer
0.63 mm
For Use With (Application)
For making microvolume injections of 1.5μL to 8μL
Length (English)
1.65 in.
Length (English) Needle
1.65 in.
Length (Metric)
42 mm
Product Type
Microvolume Manual Syringe
Volume (Metric)
10 μL
Needle Point Style
Cone
Connection Type
Fixed