Autosampler Syringes for Agilent GCs
Autosampler Syringes for Agilent GCs
Actual product may vary
Autosampler Syringes for Agilent GCs
Accurate and reproducible injections. Cone-tipped dual gauge needle. Available with or without Gas Tight plunger.
Have Questions?
Catalog NumberVolume (Metric)
SGE00282410 μL
Catalog number SGE002824
Price (MYR)
-
Volume (Metric):
10 μL
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
MaterialBorosilicate Glass (Barrel)
Needle Gauge23 to 26 G
Diameter (Metric) Inner Needle0.11 mm
Diameter (Metric) Outer6.5 mm
Diameter (Metric) Outer Needle0.63/0.47 mm
Product TypeGC Autosampler Syringe
Quantity25/Pk.
Volume (Metric)10 μL
Length (Metric) Needle42 mm
Needle Point StyleCone
For Use With (Equipment)Agilent
Connection TypeFixed
Unit SizePK