Manual Syringes For Rheodyne™ and Valco™ Valves
Manual Syringes For Rheodyne™ and Valco™ Valves
Actual product may vary
Manual Syringes For Rheodyne™ and Valco™ Valves
Manual Syringes For Rheodyne™ and Valco™ Valves
Manual Syringes For Rheodyne™ and Valco™ Valves
The injection size sets the injection volume
Have Questions?
Catalog NumberVolume (Metric)Connection TypeModel
SGE00230110 μLFixed10F-LC
Catalog number SGE002301
Price (MYR)
267.00
EA
Add to cart
Volume (Metric):
10 μL
Connection Type:
Fixed
Model:
10F-LC
Price (MYR)
267.00
EA
Add to cart
  • Gauge: 22
  • Needle Length: 5.08cm (2 in.)
  • Tip Style: LC
Specifications
Needle Gauge22 G
Diameter (English) Outer Needle0.028 in.
Diameter (Metric) Outer Needle0.71 mm
Length (English)2 in.
Length (English) Needle2 in.
Product TypeManual Syringe
Model10F-LC
QuantityEa.
Volume (Metric)10 μL
Needle Point StyleLC
For Use With (Equipment)Rheodyne and Valco Valves
Connection TypeFixed
Unit SizeEA