Gastight Syringes: Fixed Needle Models
Gastight Syringes: Fixed Needle Models
Actual product may vary
Gastight Syringes: Fixed Needle Models
Gastight Syringes: Fixed Needle Models
Gastight Syringes: Fixed Needle Models
Suitable for liquid and gas samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeModel
SGE00220010 μL26 G10F-GT
SGE00420050 μL25 G25F-GT
SGE005200100 μL25 G25F-GT
SGE006200250 μL25 G250F-GT
SGE007200500 μL25 G500F-GT
SGE0081021 mL22 G1MDF-GT
Catalog number SGE002200
Price (MYR)
267.00
EA
Add to cart
Volume (Metric):
10 μL
Needle Gauge:
26 G
Model:
10F-GT
Price (MYR)
267.00
EA
Add to cart
Suitable for liquid and gas samples
  • 10μL to 100mL capacities
  • Constructed from borosilicate glass, with PTFE plunger tips and Kel-F™ Luer cones (where applicable)
  • Needles align with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Replaceable plungers on all models
  • Fixed-needle models have needle glued into barrel with epoxy solvent-resistant glue
Specifications
Accuracy±1%
MaterialBorosilicate Glass (Barrel), PTFE (Tip)
Needle Gauge26 G
Diameter (English) Outer Needle0.01 in.
Diameter (English) Outer0.25 in.
Diameter (Metric) Outer6.5 mm
Diameter (Metric) Outer Needle0.47 mm
For Use With (Application)Suitable for gases or liquids for maximum inertness
IncludesPlunger protection, needle glued into barrel with epoxy solvent-resistant glue
Length (English) Needle1.96 in.
Product TypeGastight Syringe
Model10F-GT
QuantityEa.
Volume (Metric)10 μL
Needle Point StyleFixed Bevel
Unit SizeEA