Autosampler Syringes
Autosampler Syringes
Actual product may vary
Autosampler Syringes
Autosampler Syringes
Autosampler Syringes
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Have Questions?
Catalog NumberVolume (Metric)Needle GaugeConnection TypeFor Use With (Equipment)
SGE0019545 μL23 GFixedPerkinElmer Autosystem
Catalog number SGE001954
Price (MYR)
719.00
EA
Add to cart
Volume (Metric):
5 μL
Needle Gauge:
23 G
Connection Type:
Fixed
For Use With (Equipment):
PerkinElmer Autosystem
Price (MYR)
719.00
EA
Add to cart
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Specifications
Needle Gauge23 G
Diameter (English) Outer Needle0.02 in.
Diameter (Metric) Outer Needle0.63 mm
For Use With (Application)Gas chromatography
Length (English)2.75 in.
Length (English) Needle2.7 in.
Length (Metric)70 mm
Product TypeAutosampler Syringe
Model5F-PE-GT-0.63C
QuantityEa.
Volume (Metric)5 μL
Length (Metric) Needle70 mm
Needle Point StyleCone
For Use With (Equipment)PerkinElmer Autosystem
Connection TypeFixed
Unit SizeEA