Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 μL
Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 μL
Actual product may vary
Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 μL
Incorporate Diamond Syringe Technology offering significantly improved levels of durability, clarity and accuracy. SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle are designed specifically for use on Agilent autosamplers.
Have Questions?
Catalog NumberNeedle GaugeDiameter (Metric) Outer Needle
SGE00181423 G0.63 mm
Catalog number SGE001814
Price (MYR)
3,018.00
PK
Add to cart
Needle Gauge:
23 G
Diameter (Metric) Outer Needle:
0.63 mm
Price (MYR)
3,018.00
PK
Add to cart
  • Improved solvent resistance
  • Wide temperature ranges
  • Increased operational smoothness
  • Color coded syringes by volume
  • Range of needle gauge and length options
Specifications
DescriptionNot gas tight
Needle Gauge23 G
Diameter (Metric) Outer Needle0.63 mm
For Use With (Application)Gas chromatography
Length (Metric)42 mm
Product TypeAutosampler Syringe
Model5F-AG-0.63C 6 pack
Volume (Metric)5 μL
Needle Point StyleCone
For Use With (Equipment)Agilent GC Autosamplers 7693 A, 7673, 7683, and 6850 ALS
Connection TypeFixed
Unit SizePK