Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Actual product may vary
Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionHoldsFor Use With (Equipment)
RH32-13
also known as AVL
Glass and Plastic Tubes: 32 (13 x 100mm), Medium capacity models32 Glass and Plastic TubesSPD1030, SPD120, SPD130, SPD140DDA
RH10-15
also known as AVL
Centrifuge Tubes: 10 x 15mL conical, Medium capacity modelsCentrifuge TubesSPD1030, SPD120, SPD130, SPD140DDA
RH40-11
also known as AVL
Microtubes: 40 x 1.5mL, Medium capacity models40 MicrotubesSPD1030, SPD120, SPD130, SPD140DDA
RH60-12-40Vials: 60 (12 x 32mm), Medium capacity models60 VialsSPD1030, SPD120, SPD130, SPD140DDA
RH8-17.5Glass and Plastic Tubes: 8 (17.5 x 102mm, 15mL Corex), Medium capacity models17.5 x 102 mm, 15 mL Corex™SPD1030, SPD120, SPD130, SPD140DDA
Catalog number RH32-13
also known as AVL
Price (MYR)
6,599.00
EA
Add to cart
Description:
Glass and Plastic Tubes: 32 (13 x 100mm), Medium capacity models
Holds:
32 Glass and Plastic Tubes
For Use With (Equipment):
SPD1030, SPD120, SPD130, SPD140DDA
Price (MYR)
6,599.00
EA
Add to cart
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Various size rotors made from either plastic or aluminum to fit various sample formats of your choice.
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
Specifications
Quantity1 Ea.
DescriptionGlass and Plastic Tubes: 32 (13 x 100mm), Medium capacity models
MaterialGlass (Tube), Plastic (Tube), Glass (Tube), Plastic (Tube)
TypeRotor
For Use With (Equipment)SPD1030, SPD120, SPD130, SPD140DDA
Holds32 Glass and Plastic Tubes
Product TypeRight Analysis Spoon Spatula
ModelRH32-13
Rotor SystemTraditional
Blade MaterialSteel
Length (Metric)180 mm
Dimensions (L x W)13 x 100 mm
Unit SizeEA