Check Order Status
Rotor For Two Shallow-Well Microplates RH2MP
Rotor For Two Shallow-Well Microplates RH2MP
Rotor; Thermo Scientific; For two shallow-well microplates; Holds two plates per run; RH2MP
Have Questions?
Catalog number
RH2MP
also known as AVL
Price (MYR)
10,649.00
EA
Add to cart
Price (MYR)
10,649.00
EA
Add to cart
Rotor; Thermo Scientific; For two shallow-well microplates; Holds two plates per run; RH2MP
Specifications
Type
Concentrator Rotor
For Use With (Equipment)
Savant™ ISS110 DNA SpeedVac™ concentrator
Holds
2 Plates