Precision™ Compact Ovens
Precision™ Compact Ovens
Actual product may vary
Precision™ Compact Ovens
Conserve valuable benchtop space with these compact heating and drying ovens, available in your choice of mechanical or gravity convection models.
Have Questions?
Catalog NumberModelChamber MaterialDisplayVoltage
PR305220GGravity ConvectionAluminumAnalog240 V
Catalog number PR305220G
Price (MYR)
17,898.00
EA
Add to cart
Model:
Gravity Convection
Chamber Material:
Aluminum
Display:
Analog
Voltage:
240 V
Price (MYR)
17,898.00
EA
Add to cart
Conserve valuable benchtop space with Thermo Scientific™ Precision™ Compact Ovens, featuring mechanical or gravity convection. These compact ovens, ideal for drying and baking applications, have a double-wall interior with 2.5cm (1in.) silica-based insulation and powder-coated cold-rolled steel exterior. Pilot light visually indicates oven operation status.
Key Features
  • Double-wall interior with 2.5cm (1in.) silica-based insulation and powder-coated cold-rolled steel exterior
  • Pilot light visually indicates oven operation status
Gravity-Convection Models
  • Gentle drying with low turbulence: air is moved vertically through chamber to heat samples
  • Preset high-temperature safety bimetallic thermostat ensures overtemperature protection
  • Analog control, no LED display
Mechanical-Convection Models
  • Uniform heating, precise temperature control and fast drying
  • Hydraulic thermostat
  • LED display
  • Stainless-steel chamber
Ordering Information:
Additional shelves are sold separately.
Includes:
One fixed and two adjustable shelves.
Recommended for:
Drying and baking
Warranty: 1-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Chamber MaterialAluminum
Controller TypeBimetallic Thermostat
Capacity (Metric)48 L
Temperature (Metric)Ambient +5° to 210°C
TypeGravity Convection
DisplayAnalog
ModelGravity Convection
Dimensions (D x W x H) Interior11.5 x 16 x 16.2 in. (29 x 41 x 41 cm)
Dimensions (L x W x H)13.5 x 18.5 x 22.5 in. (34 x 47 x 57 cm)
Length (English)13.5 in.
Length (Metric)34 cm
Width (English)18.5 in.
Width (English) Interior16 in.
Width (Metric)47 cm
Width (Metric) Interior41 cm
Height (English)22.5 in.
Height (English) Interior16.2 in.
Height (Metric)57 cm
Height (Metric) Interior41 cm
Depth (English) Interior11.5 in.
Depth (Metric) Interior29 cm
Shipping Weight (English)57 lb.
Shipping Weight (Metric)26 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V