Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Actual product may vary
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Owl™ Dual-Gel Vertical Electrophoresis Systems
Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) BufferIncludes
P9DS-2550 mLComplete System
P10DS-11250 mLComplete System plus Caster
P8DS-1300 mLComplete System plus Caster
P8DS-2300 mLComplete System
P9DS-1550 mLComplete System plus Caster
Catalog number P9DS-2
Price (MYR)
14,013.00
EA
Add to cart
Volume (Metric) Buffer:
550 mL
Includes:
Complete System
Price (MYR)
14,013.00
EA
Add to cart
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel
Features:
  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
Specifications
Product LineOwl
TypeDual-Gel Vertical Electrophoresis System
IncludesComplete System
ModelP9DS
Volume (Metric) Buffer550 mL
Length (English)9.8 in.
Length (Metric) Gel14 to 16 cm
Width (English)5.1 in.
Width (Metric) Gel16 cm
Height (English)7.7 in.
Unit SizeEA
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.