Check Order Status
High-Pressure Crosses
Actual product may vary
High-Pressure Crosses
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Have Questions?
Catalog Number
Diameter (English) Inner
P-729
0.02 in.
Catalog number
P-729
Price (MYR)
-
Diameter (English) Inner:
0.02 in.
Specifications
Type
Cross
Diameter (English) Inner
0.02 in.
Pressure Rating
3500 psi
Swept Volume
0.721 μL
Material
PEEK
Color
Black
For Use With (Equipment)
1/16 in. O.D. Tubing