Heat Insulated Ceramic Pads
Heat Insulated Ceramic Pads
Actual product may vary
Heat Insulated Ceramic Pads
Keep your surfaces safe from hot items. The GSC International Heat Insulated Ceramic Pads are made from flexible alumina and silica oxides.
Have Questions?
Catalog NumberDimensions (L x W)
OPTC-P66 x 6 in.
Catalog number OPTC-P6
Price (MYR)
6,009.00
EA
Add to cart
Dimensions (L x W):
6 x 6 in.
Price (MYR)
6,009.00
EA
Add to cart
  • Keep your surfaces safe with a product that has low thermal conductivity
  • Made from flexible alumina and silica oxides with small amounts of organic binder
  • Useful for protecting surfaces while working with hot items like crucibles or flasks
Specifications
Product TypePad
Length (English)6 in.
Dimensions (L x W)6 x 6 in.
For Use With (Application)Protecting Surfaces While Working with Hot Items Like Crucibles or Flasks
Width (English)6 in.
Unit SizeEA