Steriking™ Dust-cover Bags
Steriking™ Dust-cover Bags
Actual product may vary
Steriking™ Dust-cover Bags
Designed to be used in the health care establishments to provide extra protection to the sterilized health care items against contamination.
Have Questions?
Catalog NumberDimensions (L x W)
OPTC-B5400 x 550 mm
Catalog number OPTC-B5
Price (MYR)
2,616.00
EA
Add to cart
Dimensions (L x W):
400 x 550 mm
Price (MYR)
2,616.00
EA
Add to cart
The STERIKING™ dust cover bags have been designed to be used in the health care establishments to provide extra protection to the sterilized health care items against contamination. It becomes necessary when extended sterility time is needed or the items become exposed to the conditions where the hygienic requirements are not fulfilled, for example during outdoors transportation. A dust cover bag will protect a product in its directly enclosing packaging from dust and environmental influences during transport or storage. A dust cover bag is used to extend the shelf life (in respect of the sterility) of packed sterile medical devices
  • Conformity to International Standards - The STERIKING™ Dust cover bags conform to the international product standards and norms: ISO 11607-1:2019/Amd 1:2014 and ISO 11607-2:2019/Amd 1:2014.
  • The STERIKING™ Dust cover bags are constructed of a multiply BoPET/PE plastic laminate (12/50 microns). Dust covers are a peel-open bag providing superior opening characteristics compared to the conventional tear-off bags.
  • Bags are heat sealable with an impulse or rotosealer. Recommended sealing temperature for final closing is 130-160° C (272-335° F) depending on pressure and time.
  • Recommended shelf life – 10 years pre-sterilisation
Specifications
Dimensions (L x W)400 x 550 mm
For Use With (Application)Used in The Health Care Establishments to Provide Extra Protection to The Sterilized Health Care Items Against Contamination
Length (Metric)400 mm
Quantity100/Pk.
Width (Metric) Exterior550 mm
Unit SizeEA