Kemtec™ Accelerated Motion Kit
Kemtec™ Accelerated Motion Kit
Actual product may vary
Kemtec™ Accelerated Motion Kit
Investigate accelerated motion using a totally reusable system that includes six foot tubular ramps, toy cars and ball bearings.
Have Questions?
Catalog NumberGrade
OPT151048 to 12
Catalog number OPT15104
Price (MYR)
104.00
EA
Add to cart
Grade:
8 to 12
Price (MYR)
104.00
EA
Add to cart
  • Simple apparatus with an exceptional support system that allows students to use timed trials to measure gravitational acceleration and the car's/ball's coefficient of friction
  • Adjustable system allows modification of track length and angle
  • Includes separate problem sets for general science and higher level Physics courses
  • Higher level curriculum includes kinetic and static coefficients of friction as well as bodies on inclined planes
  • Instructor's manual includes reproducible student instructions, guided calculations including graphing of data and calculation of error, assessment questions, sample data and answer keys
  • Appropriate for any level Physics course (including AP) as well as general science courses
  • Grades 9 to 12
  • Class size: 24 students
Specifications
Product TypeAccelerated Motion Kit
Grade8 to 12
For Use With (Application)Engages students in study of the basic laws of physics, kit investigates that phenomenon and relates it to a vector component of the acceleration due to gravity
IncludesInstructor manual, student data sheet, pvc couplers, section of pvc pipe, 3-prong clamp extensions, right angle clamp, stopwatch, die cast car, metric tape, protractor, steel ball bearing, graph paper
Class Size36 Students
Unit SizeEA