Prefilled 150 mL Formalin Container
Prefilled 150 mL Formalin Container
Actual product may vary
Prefilled 150 mL Formalin Container
Protects against exposure to harmful chemicals and vapors
Have Questions?
Catalog NumberColor
OPT15008Lilac
Catalog number OPT15008
Price (MYR)
-
Color:
Lilac
  • Reduces or eliminates exposure to formalin
  • Reduces exposure to harmful vapors
  • Comes in a variety of colored caps for easy specimen identification
  • Security seal
  • Dual Chamber keeps formalin trapped until container is activated
Specifications
SterilityNon-sterile
Capacity (Metric)150 mL
Closure MaterialPolypropylene
Closure TypeThreaded Cap
ColorLilac
For Use With (Application)Pathology Specimens
Height (English)3.625 in.
No. per Case96/Cs.
No. per Pack24/Pk.
Width (English)2.25 in.
Unit SizeEA