Density Determination Kit for Balances
Density Determination Kit for Balances
Actual product may vary
Density Determination Kit for Balances
Density Determination Kit for Balances
Density Determination Kit for Balances
Use with select OHAUS™ Balances with 0.1 to 1mg resolutions.
Have Questions?
Catalog number OHA80253384
Price (MYR)
5,255.00
EA
Add to cart
Price (MYR)
5,255.00
EA
Add to cart
Compatible with:
Models with capacities up to 610g and readability of 0.1 and 1mg
  • Discovery
  • Explorer
  • Adventurer Pro (except AV53x)
  • Pioneer
Models with capacities up to 1100g and readability of 0.1 and 1mg
  • Explorer
Specifications
For Use With (Equipment)Ohaus Balance
IncludesWeighing platform, holder for floating items, hold
LanguagesChinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Product LineDensity Determination Kits
TypeDensity Determination Kit
Unit SizeEA