MB25 Moisture Analyzer
MB25 Moisture Analyzer
Actual product may vary
MB25 Moisture Analyzer
Provides precise moisture content determination, percent solid determination and accurate weighing at an unbeatable value. Ohaus™ MB25 Moisture Analyzer delivers the ideal combination of results and value.
Have Questions?
Catalog number OHA80252472
Price (MYR)
-
  • Large backlit LCD display
  • Standard RS232 port
  • Power cord included
  • 110g capacity
  • Readability of 0.005g/0.05%
  • Halogen heating
  • ABS housing, metal pan support, metal pan handler, in-use cover
  • 3 shut off criteria (manual, timed or auto), 1 standard heating profile, 50° to 160°C heating range (5°C increments)
Specifications
ModelMB25
MaterialABS Housing, Metal Pan Support, Metal Pan Handler
Dimensions (L x W x H)280.00 x 165.00 x 127.000 mm
Display TypeDigital LCD
Connection TypeRS232 (Included)
Depth (Metric)280.00 mm
DescriptionMoisture Analyzer, MB25
For Use With (Application)In a Contract Laboratory, Wastewater Treatment Pla
Height (English)5.00 in.
Height (Metric)127.000 mm
Length (English)11.00 in.
Length (Metric)280.00 mm
Net Weight (English)5.000 lb.
Net Weight (Metric)2.100 kg
Temperature Range (Metric)50°C to 160°C,
TypeMoisture Analyzer
Width (English)6.50 in.
Width (Metric)165.00 mm
Unit SizeEA