Vortex Mixer
Vortex Mixer
Actual product may vary
Vortex Mixer
Powerful Hands-Free Vortex Mixer for Efficient High-Throughput Mixing
Have Questions?
Catalog NumberController Type
OHA30573837Digital
Catalog number OHA30573837
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Controller Type:
Digital
  • Easily process up to 50 samples at one time - the Multi-Tube Vortex Mixer can be customized for your application with six different tube racks and a post- extension kit.
  • The Multi-Tube Vortex Mixers feature either an easy to clean one-piece stainless steel housing that is perfect for clean rooms or a painted steel version for a more economical option.
  • For challenging samples, enhance vortex action & create a more vigorous mix with the programmable pulse mode on the digital models. Adjust the pulse-on/pulse-off time interval to customize operation
Specifications
Orbit3.6 mm
Voltage230 V
Wattage100 W
Frequency50/60 Hz
MaterialStainless Steel foam rack for (50) 12 mm diameter tubes (included), interchangeable foam racks for vortexing tubes from 10 - 29 mm in diameter
Dimensions (L x W)241 x 384 mm
Height (English)16 in.
Height (Metric)406 mm
Length (English)9.5 in.
Width (English)15.1 in.
Shipping Weight (Metric)19.1 kg
Weight (English)41 lb.
Weight (Metric)18.7 kg
Controller TypeDigital
ModelVXMTDGB
DisplayIndependent LED displays for speed/time
Unit SizeEA