Check Order Status
DDR2-NanoLuc Fusion Vector, 20microg
Actual product may vary
DDR2-NanoLuc Fusion Vector, 20microg
Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.
Have Questions?
Catalog Number
Quantity
OHA30467932
20 μg
Catalog number
OHA30467932
Price (MYR)
1,233.00
EA
Add to cart
Quantity:
20 μg
Price (MYR)
1,233.00
EA
Add to cart
Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
Blue/white screening for easy identification of recombinant clones
In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
The difference between + and − vectors is orientation of the f1 origin
Specifications
Quantity
20 μg
Storage Requirements
-30°C to -10°C