Check Order Status
Alarm Beacon Kit
Actual product may vary
Alarm Beacon Kit
OHAUS™ Alarm Beacon Kit is designed to be used with a bench scale, indicator, or manual light tower.
Have Questions?
Catalog number
OHA30424021
Price (MYR)
-
For use with all OHAUS™ Bench Scales and Indicators
Can be used with manual light tower
Specifications
Color
Red, Orange, Green
Height (Metric)
243 mm
Languages
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Weight (Metric)
210 g
Type
Column Kit