Check Order Status
MB27 Basic Moisture Analyzer
Actual product may vary
MB27 Basic Moisture Analyzer
OHAUS™ MB27 Basic Moisture Analyzer combines high quality and durable construction into a sleek, compact design.
Have Questions?
Catalog number
OHA30251541
Price (MYR)
-
With 0.01% 0.001g accuracy, the MB27 Moisture Analyzer is suitable for most applications.
Halogen heating and dual weight-temperature calibration provide fast, reliable results.
User-friendly touch-button design for easy operation.
Specifications
Model
MB27
Dimensions (L x W x H)
280.00 x 165.00 x 127.000 mm
Display Type
LCD
Connection Type
RS232 (included)
Capacity
90 g
Net Weight (Metric)
0.177 kg
Weight (Metric)
2.800 kg