Starter Series Electrode: STSURF 2-in-1 Flat pH Electrode
Starter Series Electrode: STSURF 2-in-1 Flat pH Electrode
Actual product may vary
Starter Series Electrode: STSURF 2-in-1 Flat pH Electrode
Use in combination with Starter bench and portable meters. OHAUS Starter Series Electrode: STSURF 2-in-1 Flat pH Electrodes need to work with STTEMP30 temperature electrodes for 0.01 pH accuracy.
Have Questions?
Catalog NumberConnector Types
OHA30129470BNC
Catalog number OHA30129470
Price (MYR)
796.00
EA
Add to cart
Connector Types:
BNC
Price (MYR)
796.00
EA
Add to cart
  • Designed to produce exact results time and time again
  • Sturdy and durable
  • Constructed from plastic shafts and built to withstand daily use
  • Includes a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements
  • Fits in electrode holders on bench meters and electrode clips on all portable meters
  • Reference Electrolyte: 3M KCl Solution
Specifications
  • Parameters Measured: pH
  • pH Range: 0.00 to 14.00
  • Temperature Range: 0° to 80°C
  • Connection: BNC
  • Cable Length: 1 m
  • Shaft dia. x L: 12 x 120 mm
  • Zero Potential: 7.00±0.25 pH
  • Slope: ≥97%
  • Impedance: ≤250MOhm
  • Reference Diaphragm: Sleeve
  • Application Scope: surface, skin, leather, paper, emulsion
Applications
Ideal for use in lab or in the field.
Specifications
Connector TypesBNC
JunctionGlass Fiber
MaterialPlastic
pH Range0 to 14
Temperature (Metric) Operating0 to 100°
Type2-in-1 pH Electrode
Filling Solution Type3M KCl Solution
For Use With (Equipment)Starter bench and Portable meters
ModelSTSURF
Temperature Accuracy0 to 80°C
Length (Metric) Cable1 m
Length (Metric) Stem120 mm
Diameter (Metric) Body12 mm
Unit SizeEA