Human IFN gamma ELISA Development Kit (TMB), PeproTech®
Human IFN gamma ELISA Development Kit (TMB), PeproTech®
Actual product may vary
Human IFN gamma ELISA Development Kit (TMB), PeproTech®
Human IFN gamma ELISA Development Kit (TMB), PeproTech®
Human IFN gamma ELISA Development Kit (TMB), PeproTech®
ELISA
Have Questions?
Catalog NumberQuantity
OHA3008799710 x 96 Tests
Catalog number OHA30087997
Price (MYR)
348.00
EA
Add to cart
Quantity:
10 x 96 Tests
Price (MYR)
348.00
EA
Add to cart
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
Specifications
Accession NumberP01579
Assay RangeLot-specific
Assay SensitivityLot-specific
ConjugateHRP
Product TypeELISA
For Use With (Equipment)Colorimetric Microplate Reader
Gene ID (Entrez)3458
Gene SymbolIFNG
Kit ContentsCapture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
Label or DyeHRP
Quantity10 x 96 Tests
Regulatory StatusRUO
Storage RequirementsRefer to product documentation for component specific storage temperature.
TargetIFN gamma
Test Time1 hrs 45 mins
Total Assay Time24 hrs 30 mins
Unit SizeEA