Starter Conductivity Pen Meters With Temperature Display
Starter Conductivity Pen Meters With Temperature Display
Actual product may vary
Starter Conductivity Pen Meters With Temperature Display
Small but productive conductivity meters. OHAUS Starter Conductivity Pen Meters with Temperature Display are the choice for basic testing needs of the total ionic concentration within a solution. They are durable with IP67 waterproof design and provide fast and straight forward results.
Have Questions?
Catalog NumberDisplay Type
OHA30073975LCD
Catalog number OHA30073975
Price (MYR)
1,334.00
EA
Add to cart
Display Type:
LCD
Price (MYR)
1,334.00
EA
Add to cart
Models ST20C-C, ST20C-B, and ST20C-A, come with a temperature display.
  • Equipped with wrist strap for security
  • Accuracy: ±1.5% FS
  • Dual Display with temperature Display
  • Automatic Shutdown after 6 minutes of Non-Use
  • Requires 4 AG13 1.5V Micro Alkaline Batteries
  • Size/Weight: 185 × 45 × 38 mm, 110 g
Specifications
TypeConductivity Pen Meter
Display TypeLCD
Range (Conductivity)0 to 19.99 μS/cm
Resolution0.1 μS/cm
Accuracy±1.5% FS
Ambient Temperature (English)32 to 122°F
Ambient Temperature (Metric)0 to 50°C
Battery TypeMicro Alkaline
MaterialABS
Temperature (English) Operating32 to 211°F
Temperature (Metric) Operating0 to 99°C
Weight (English)0.24 lb.
Weight (Metric)110 g
For Use With (Application)For basic testing of the total ionic concentration within a solution
Unit SizeEA