Explorer™ Precision Balances
Explorer™ Precision Balances
Actual product may vary
Explorer™ Precision Balances
The Explorer has risen to the next level of ingenuity; expanding on its modern features and design to offer unmatched functionality.
Have Questions?
Catalog NumberReadabilityModelStabilization TimeSample Size
OHA300619810.01 gEX2202/E≤1 sec.2.2 kg
Catalog number OHA30061981
Price (MYR)
23,472.00
EA
Add to cart
Readability:
0.01 g
Model:
EX2202/E
Stabilization Time:
≤1 sec.
Sample Size:
2.2 kg
Price (MYR)
23,472.00
EA
Add to cart
  • Large color touch-screen display
  • Icon-driven application software that features 14 unique application modes
  • 3 level adjustable angle display
  • Newly optimized linearity and repeatability specifications
  • Enhanced vibration filtering for better balance stability
  • All models come standard with AutoCal™ automatic internal calibration
  • 3 x 3 filtering setup ensures fast and stable weighing results
  • The base and display separate for maximum flexibility in anywork area
  • Draftshield models isolate the weighing chamber to enhance stability and speed results while offering maximum accessibility to the weighing chamber through top-mounted side doors or through the versatile top door that flips or slides open
  • Hands-free entry is available with optional automated side doors
  • Easy access communication ports (USB, RS232) plus 3rd communication port slot (Ethernet or 2nd RS232)
  • GLP and GMP including date and time, Direct Data Transfer
  • Metal base, ABS top housing, stainless steel pan, glass draftshield with flip-top door, replaceable in-use cover
  • 3-door draftshield with removable panels on 0.1mg and 1 mg models with draftshield light
  • Modular display design with adjustable angle (attach to base, wall or optional tower)
  • AutoCal automatic internal calibration
  • Level assist feature with illuminated level indicator
  • Selectable environmental and auto-print settings
  • Integral weigh belowhook
  • Protective in-use cover
  • Software lock switch
  • High resolution 14.5cm (diagonal) full colour VGA touch display including QWERTY and numeric key pads plus weighing capacity indication
Specifications
Automation InterfaceTouch screen
Certifications/ComplianceNIST, AM-5847, IEC/EN 61010-1:2010, CAN/CSA-C22.2,
Connection TypeRS-232, USB, Optional Ethernet or 2nd RS232
Capacity2200 g
Display TypeColor Graphic
Readability0.01 g
FormatGram, milligram, kilogram, carat, ounce, troy ounc
IncludesAutomatic draft shield 3 door, internal (AutoCal)
Linearity±0.02 g
MaterialABS Plastic, Stainless Steel, Glass
ModelEX2202/E
Repeatability<±0.01 g
Sample Size2.2 kg
Sensitivity Drift1.5ppm/°C
Stabilization Time≤1 sec.
Temperature Range (English)50°F to 86°F
Temperature Range (Metric)10°C to 30°C
TypePrecision Balance
Width (English)9.1 in.
Width (Metric)23 cm
Height (English)3.9 in.
Height (Metric)9.8 cm
Depth (English)15.5 in.
Depth (Metric)39.3 cm
Weight (English)9.5 lb.
Weight (Metric)4.3 kg
Electrical Requirements100/240 V 50/60 Hz
Voltage100/240 V
Amperage0.6 A