NM23-H2/NME2 Antibody (NME2/6437) [Janelia Fluor™ 646], Novus Biologicals™
NM23-H2/NME2 Antibody (NME2/6437) [Janelia Fluor™ 646], Novus Biologicals™
Actual product may vary
NM23-H2/NME2 Antibody (NME2/6437) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Have Questions?
Catalog number OHA30060845
Price (MYR)
-
NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
AntigenNM23-H2/NME2
ApplicationsWestern Blot, Immunohistochemistry (Paraffin)
ClassificationMonoclonal
CloneNME2/6437
ConjugateJanelia Fluor 646
Formulation50 mM Sodium Borate
Gene AliasEC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in
Host SpeciesMouse
ImmunogenRecombinant full-length human NM23-H2/NME2 protein
Purification MethodProtein A or G purified
Quantity0.1 mL
Regulatory StatusRUO
Research DisciplineCell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or SecondaryPrimary
Gene ID (Entrez)4831
Target SpeciesHuman
FormPurified
IsotypeIgG1 κ
Unit SizeEA
Contents & Storage
Store at 4°C in the dark.