TAFA3/FAM19A3 Antibody (460904), Alexa Fluor™ 488, Novus Biologicals™
TAFA3/FAM19A3 Antibody (460904), Alexa Fluor™ 488, Novus Biologicals™
Actual product may vary
TAFA3/FAM19A3 Antibody (460904), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
OHA300592560.1 mL
Catalog number OHA30059256
Price (MYR)
303.00
EA
Add to cart
Quantity:
0.1 mL
Price (MYR)
303.00
EA
Add to cart
TAFA3/FAM19A3 Monoclonal antibody specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
AntigenTAFA3/FAM19A3
ApplicationsWestern Blot, Flow Cytometry, CyTOF
ClassificationMonoclonal
Clone460904
ConjugateAlexa Fluor 488
DilutionWestern Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation50mM Sodium Borate
Gene AliasChemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3
Host SpeciesMouse
ImmunogenE. coli-derived recombinant human TAFA3/FAM19A3, Ala31-Arg133, Accession # Q7Z5A8
Purification MethodProtein A or G purified from hybridoma culture supernatant
Quantity0.1 mL
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)284467
Target SpeciesHuman
FormPurified
IsotypeIgG2b
Unit SizeEA
Contents & Storage
Store at 4C in the dark.