Mouse Serpin F1/PEDF ELISA Kit (Colorimetric)
Mouse Serpin F1/PEDF ELISA Kit (Colorimetric)
Actual product may vary
Mouse Serpin F1/PEDF ELISA Kit (Colorimetric)
Have Questions?
Catalog NumberQuantity
OHA300592551 Kit
Catalog number OHA30059255
Price (MYR)
303.00
EA
Add to cart
Quantity:
1 Kit
Price (MYR)
303.00
EA
Add to cart
Specifications
Assay Range0.16 to 10 ng/mL
Assay Sensitivity0.09 ng/mL
Product TypeELISA Kit (Colorimetric)
Sample TypeSerum, Plasma, Other Biological Fluids
SpecificityThis kit recognizes Mouse Serpin F1/PEDF in samples. No significant cross-reactivity or interference between Mouse Serpin F1/PEDF and analogues was observed.
For Use With (Application)ELISA, Sandwich ELISA
Gene ID (Entrez)5176
Gene SymbolSERPINF1
Quantity1 Kit
Research DisciplineBiologically Active Proteins, Neuroscience, Vision
Storage RequirementsStorage of components varies. See protocol for specific instructions.
SynonymCell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDF (pigment epithelium-derived factor, pigment epitheliumderived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium-derived factor), member 1
TargetMouse
Unit SizeEA