NanoDrop™ One/OneC Productivity Kits
NanoDrop&trade; One/One<sup>C</sup> Productivity Kits
Actual product may vary
NanoDrop™ One/OneC Productivity Kits
The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.
Have Questions?
Catalog NumberType
ND-PP1NanoDrop One Productivity Kit
Catalog number ND-PP1
Price (MYR)
6,353.00
EA
Add to cart
Type:
NanoDrop One Productivity Kit
Price (MYR)
6,353.00
EA
Add to cart
The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.
The NanoDrop One Productivity Kit includes:
  • NanoDrop-branded Finnpipette™ F1 Adjustable-Volume Pipette , 0.2–2 μL
  • Microfiber screen wipe for NanoDrop One Spectrophotometer
  • NanoDrop PR-1 Reconditioning Compound Kit
  • NanoDrop PV-1 Performance Verification Solution
  • I love my NanoDrop sticker
The NanoDrop OneC Productivity Kit includes:
  • NanoDrop-branded Finnpipette™ F1 Adjustable Volume Pipette , 0.2–2μL
  • Microfiber screen wipe for NanoDrop OneC spectrophotometer
  • NanoDrop PR-1 Reconditioning Compound Kit
  • NanoDrop PV-1 Performance Verification Solution
  • 10mm path quartz cuvette
  • Cuvette stir bars, 5x2mm, pack of 10
  • I love my NanoDrop sticker
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeNanoDrop One Productivity Kit
For Use With (Equipment)NanoDrop One
Includes
  • NanoDrop-branded Finnpipette™ F1 Adjustable-Volume Pipette , 0.2–2 μL
  • Finntip™ Pipette Tips, 10 μL , sterile, pack of 96
  • Microfiber screen wipe for NanoDrop One spectrophotometer
  • NanoDrop One spectrophotometer USB memory device
  • NanoDrop PR-1 Reconditioning Compound Kit
  • NanoDrop PV-1 Performance Verification Solution
  • “I love my NanoDrop” sticker
Storage RequirementsStore at room temperature
Unit SizeEA