Muffle Furnaces
Muffle Furnaces
Actual product may vary
Muffle Furnaces
Nabertherm furnaces manufactured from expertly finished, high quality materials and are easy to operate. The insulation out of non-classified fibre materials permits energy saving operation and fast heating times due to low heat storage and thermal conductivity.
Have Questions?
Catalog NumberCapacity (Metric)Controller TypeStyleTemperature (Metric)
NABL-154V4LN1115 LB510Lift Door1400°C
Catalog number NABL-154V4LN11
Price (MYR)
-
Capacity (Metric):
15 L
Controller Type:
B510
Style:
Lift Door
Temperature (Metric):
1400°C
  • Heating from two sides by ceramic heating plates with integrated heating element which is safeguarded against fumes and splashing, and easy to replace
  • Insulation made of non-classified fibre material
  • Textured stainless steel sheet housing
  • Equipped with either a flap door (L) or lift door (LT)
  • Dual shell housing for low external temperatures and high stability
  • Adjustable air inlet integrated into the door, exhaust air outlet in the rear wall of furnace
  • Supplied with B510 controller with four programs and five freely programmable ramps, or C550 with 10 freely adjustable programs and 20 segments
  • Both controllers have integrated USB interface and offer plain text display in different languages
  • Alternative furnace sizes and temperature ranges are available on request
Specifications
Controller TypeB510
Capacity (Metric)15 L
Temperature (Metric)1400°C
TypeFurnace
StyleLift Door
Dimensions (D x W x H) Interior350 x 250 x 170 mm
Dimensions (L x W x H) Exterior625 x 530 x 980 mm
Voltage230 V
Wattage3500 W
Unit SizeEA