Specially insulated gloves for working with hot mechanical components. Nabertherm™ Carbon Fiber Gloves are heat resistant up to 650°C.
Have Questions?
Catalog Number
Temperature (Metric) Max
NAB493000004
650°C
Catalog number NAB493000004
Price (MYR)
1,403.00
EA
Add to cart
Temperature (Metric) Max:
650°C
Price (MYR)
1,403.00
EA
Add to cart
Five-fingered, full glove
Carbon fiber reinforced
Highly heat resistant
Short-time contact temperature: 650°C
Note regarding Max Temperature: The contact heat test of a glove tests its insulation capacity. It determines the maximum temperature at which the glove will protect for 15 seconds without the temperature inside the glove rising by 10 °C. Level 4 is the highest performance level. The test is carried out at room temperature. The glove is in constant contact with the hot object.