Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Product Image
Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
N10801Wash Head (1 x 12)Wellwash Microplate Strip Washer
N10805Wash BottleWellwash Microplate Strip Washer
N10800Wash Head (1 x 8)Wellwash Microplate Strip Washer
N10814Waste BottleWellwash Versa Microplate Strip Washer
Catalog number N10801
Price (MYR)
6,219.00
EA
Add to cart
Type:
Wash Head (1 x 12)
For Use With (Equipment):
Wellwash Microplate Strip Washer
Price (MYR)
6,219.00
EA
Add to cart
These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.
Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
Specifications
TypeWash Head (1 x 12)
For Use With (Equipment)Wellwash Microplate Strip Washer
Unit SizeEA