Microplate Reader Verification Plates
Microplate Reader Verification Plates
Actual product may vary
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
N03394Spectrophotometric Verification Plate
24072800Multiskan Verification Plate, 8 wavelengths
Catalog number N03394
Price (MYR)
20,304.00
EA
Add to cart
Product Type:
Spectrophotometric Verification Plate
Price (MYR)
20,304.00
EA
Add to cart
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation. 
Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeSpectrophotometric Verification Plate
For Use With (Equipment)Varioskan LUX Microplate Reader, Multiskan Sky, Multiskan Skyhigh
Plate TypeVerification Plate
Product LineVarioskan™
IncludesNPL (UK National Physical Laboratory) and NIST (US National Institute for Standards and Technology) traceable Spectrophotometric Verification Plate
Certificate of Calibration
Adapter for Spectrophotometric Verification Plate
USB memory device including SkanIt™ software sessions and reference files
Carrying case for Spectrophotometric Verification Plate
Unit SizeEA