T-Mop™ Irradiated Covers
T-Mop™ Irradiated Covers
Actual product may vary
T-Mop™ Irradiated Covers
A range of irradiated T-mop covers with snaps that fit onto the 180° swivel TMop™ Adapter to effortlessly clean and sanitize cleanroom surfaces, including soft walls, awkward corners, tight spaces and under equipment.​
Have Questions?
Catalog NumberMaterialDimensionsIncludes
MICT6-14IRSmooth Knit Polyester, Urethane Foam35.6 x 6.4 cm (14 x 2.5 in.)NovaPoly Slipcover, Irradiated
Catalog number MICT6-14IR
Price (MYR)
2,390.00
CS
Add to cart
Material:
Smooth Knit Polyester, Urethane Foam
Dimensions:
35.6 x 6.4 cm (14 x 2.5 in.)
Includes:
NovaPoly Slipcover, Irradiated
Price (MYR)
2,390.00
CS
Add to cart
  • Wide surface area for cleaning walls and ceilings
  • Flat head mop
  • Rotates 180°
  • Cleans under equipment and reaches awkward angles on shelving and narrow chambers
  • Polyester
  • Irradiated
  • White
  • Width 254mm
The Micronova Irradiated TMop™ Cleanroom Mop Covers are rounded, covered sponges with snaps that fit over the 180° swivel TMop™ Adapter to easily roll down walls and soft walls, as well as, clean awkward corners, tight spaces and under equipment. The mops are available in either textured, or smooth knit polyester fabric with option for a thicker foam layer to hold more disinfectant and facilitate wringing.
  • Irradiated T-Mop Covers with snap fasteners
  • Rounded covered sponges easily roll down walls and soft walls
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces and under equipment
  • Fabric options include:
    • NovaPoly, 100% Smooth Knit Polyester, provides excellent disinfectant dispersal
    • PolySorb, 100% Textured Polyester, assists in removing residues
  • Available with extra thick foam
  • For use with the 180° swivel TMop™ Adapter
  • Available in 10, 14 and 18-inch lengths
  • Gamma irradiated for aseptic areas and equipment
  • Holds up to strong disinfectants
  • Autoclavable
Specifications
Dimensions35.6 x 6.4 cm (14 x 2.5 in.)
Product TypeCleanroom Mop
ColorWhite
Length (English)14 in.
SterilitySterile
Length (Metric)35.6 cm
AutoclavableAutoclavable
Width (Metric)6.4 cm
Width (English)2.5 in.
Resistant ToChemicals
DisposableReusable
Quantity12/Cs.
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
IncludesNovaPoly Slipcover, Irradiated
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
MaterialSmooth Knit Polyester, Urethane Foam
Unit SizeCS