TMop™ Adapters
TMop™ Adapters
Actual product may vary
TMop™ Adapters
A selection of T-shaped mop adapters that work with TMop™ and Slim-T™ series slipcovers and feature a 180°swivel joint for easy surface cleaning in awkward corners, tight spaces and under equipment. Available in either electropolished stainless steel tube bar or acetal square bar (QDST).
Have Questions?
Catalog NumberMaterialDimensionsProduct Type
MICQPT-14Electropolished Stainless Steel (304), Acetal35.6 x 2.5 cm (14 x 1 in.)Adapter, ACME Threaded Handle Attachment
Catalog number MICQPT-14
Price (MYR)
1,087.00
EA
Add to cart
Material:
Electropolished Stainless Steel (304), Acetal
Dimensions:
35.6 x 2.5 cm (14 x 1 in.)
Product Type:
Adapter, ACME Threaded Handle Attachment
Price (MYR)
1,087.00
EA
Add to cart
The Micronova TMop™ Cleanroom Mop Adapters are designed to work with the TMop™ or Slim-T™ series mop covers that snap easily onto the T-shaped mop frames. The acetal square bar frame is designed for use with ‘thick sponge’ T-Mops, as it grips into the sponge. T-Mop™ frames are available with an ACME threaded end for plastic handles (QPT) or a Quick Disconnect attachment for use with SSU series stainless steel handles (QDT and QDST).
  • T-shaped mop adapters with 180° swivel joint
  • Easy surface cleaning in awkward corners, tight spaces and under equipment
  • Works well on room floors, walls, ceilings, soft walls and equipment exteriors
  • 304 Electropolished stainless-steel tube frame fits all TMop™ and Slim-T™Mop covers (QDT, QPT)
  • Square acetal bar works best with the ‘thick sponge’ TMop™covers (QDST)
  • QP, ACME threaded end for attachment to Micronova plastic handles
  • QD, Quick Disconnect attachment to SSU series stainless handles
  • Good resistance to cleaning solutions and disinfectants
  • Autoclavable
Specifications
Dimensions35.6 x 2.5 cm (14 x 1 in.)
Product TypeAdapter, ACME Threaded Handle Attachment
Length (English)14 in.
SterilityNon-sterile
Length (Metric)35.6 cm
AutoclavableAutoclavable
Width (Metric)2.5 cm
Width (English)1 in.
Resistant ToChemicals
DisposableReusable
Quantity1/Pk.
Care InstructionsStore at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Application)Cleaning and Sanitizing Cleanroom and Controlled Environment Surfaces
MaterialElectropolished Stainless Steel (304), Acetal
Unit SizeEA