Cleanroom Ballpoint Click Pen
Cleanroom Ballpoint Click Pen
Actual product may vary
Cleanroom Ballpoint Click Pen
Cleanroom Click Pens use low sodium, indelible ink and feature a special high-density poly barrel and easy-click retractable nib.
Have Questions?
Catalog NumberColorSterility
MICPEN-12BlueNon-sterile
Catalog number MICPEN-12
Price (MYR)
224.00
PK
Add to cart
Color:
Blue
Sterility:
Non-sterile
Price (MYR)
224.00
PK
Add to cart
  • Product Type: Ballpoint Pen
  • Ink Type: Low Sodium (Indelible)
  • Tip Type: Click / Retractible
  • Tip Style: Ballpoint (0.7 mm)
  • Dimensions (L x W): 15 cm (6 in.) x 9.5 mm (0.38 in.)
  • Packaging: Double-Bagged
  • For Use With (Application): Cleanroom Documentation
  • For Use With (Equipment): Cleanroom Paper
Specifications
ColorBlue
Cover ColorWhite
Cover MaterialHigh-Density Poly
Dimensions (L x W)15 cm (6 in.) x 9.5 mm (0.38 in.)
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 Certified Facility
DisposableReusable
For Use With (Application)Cleanroom Documentation
For Use With (Equipment)Cleanroom Paper
Packaging10 Pens/Bag, 10 Pens/Double Bag, 10 Pens/Pk.
Product TypeBallpoint Click Pen
SterilityNon-sterile
Tip StyleBallpoint (0.7 mm)
Weight (English)2.171 oz. (Pack)
Weight (Metric)61.547 g
Unit SizePK