Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Actual product may vary
Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Surface Treated SterileTissue Culture Plates
Ideal for comparison and analysis of cell growth, yield in multiple platforms.
Have Questions?
Catalog NumberCulture Area
LSVFB0129279.6 cm2
Catalog number LSVFB012927
Price (MYR)
445.00
Special offer
Ends: 31-Mar-2026
551.00
Save 106.00 (19%)
Case
Add to cart
Culture Area:
9.6 cm2
Price (MYR)
445.00
Special offer
Ends: 31-Mar-2026
551.00
Save 106.00 (19%)
Case
Add to cart
  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Specifications
Array2 x 3
Certifications/ComplianceCE
Bottom ShapeFlat
Capacity (Metric)17 mL
ColorNatural
Culture Area9.6 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentStandard
TypeCell Culture Plate
Well Count6
For Use With (Application)Tissue culture
No. per Pack100 Plates
PackagingIndividually wrapped
Unit SizeCase