Search Thermo Fisher Scientific
| Catalog Number | Diameter (Metric) | Culture Area |
|---|---|---|
| LSVFB012925 | 150 mm | 143 sq. cm |
| FB012925 | 150 mm | 143 sq. cm |
| LSVFB012920 | 35 mm | 8.5 sq. cm |
| FB012921 | 60 mm | 21.2 sq. cm |
| LSVFB012921 | 60 mm | 21.2 sq. cm |
| FB012924 | 100 mm | 60.8 sq. cm |
| LSVFB012924 | 100 mm | 60.8 sq. cm |