N-Heptane, Certified AR for Analysis, Fisher Chemical™
N-Heptane, Certified AR for Analysis, Fisher Chemical™
N-Heptane, Certified AR for Analysis, Fisher Chemical™
N-Heptane, Certified AR for Analysis, Fisher Chemical™
N-Heptane, Certified AR for Analysis, Fisher Chemical™
C7H16, CAS Number-142-82-5, dipropylmethane, dipropyl methane, eptani, n-heptane, gettysolve-c, heptan polish, skellysolve c, heptyl hydride, heptan, heptanen, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
5 L
Packaging:
View all
Amber Glass Bottle
Catalog number LSMH016017
also known as LSMH/0160/17
Price (MYR)
142.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
142.00
EA
Add to cart
Specifications
Melting Point-91°C
CAS142-82-5
ColorColorless
Boiling Point98°C
Quantity2.5 L
View more
C7H16, CAS Number-142-82-5, dipropylmethane, dipropyl methane, eptani, n-heptane, gettysolve-c, heptan polish, skellysolve c, heptyl hydride, heptan, heptanen, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900