Ethanol Absolute, for HPLC, Fisher Chemical™
Ethanol Absolute, for HPLC, Fisher Chemical™
Actual product may vary
Ethanol Absolute, for HPLC, Fisher Chemical™
Ethanol Absolute, for HPLC, Fisher Chemical™
Ethanol Absolute, for HPLC, Fisher Chemical™
C2H6O, CAS Number-64-17-5, alcohol, algrain, alcohol, anhydrol, ethyl alcohol, ethyl hydrate, ethyl hydroxide, grain alcohol, methylcarbinol, tecsol, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702
Have Questions?
Catalog number LSME0665DF17
also known as LSME/0665DF/17
Price (MYR)
1,084.00
EA
Add to cart
Price (MYR)
1,084.00
EA
Add to cart
C2H6O, CAS Number-64-17-5, alcohol, algrain, alcohol, anhydrol, ethyl alcohol, ethyl hydrate, ethyl hydroxide, grain alcohol, methylcarbinol, tecsol, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702
Specifications
pH7
Melting Point-114°C
CAS64-17-5
ColorColorless
Boiling Point78°C
Quantity2.5 L
PackagingAmber Glass Bottle
Physical FormLiquid
Molecular FormulaC2H6O
MDL Number3568
Synonymethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
IUPAC Nameethanol
Molecular Weight (g/mol)46.069
PubChem CID702
ChEBICHEBI:16236
Formula Weight46.07g/mol
Unit SizeEA