Baricitinib
Baricitinib
Actual product may vary
Baricitinib
Baricitinib
Baricitinib
Highly potent JAK1 and JAK2 inhibitor; also inhibits JAK3 and Tyk2
Have Questions?
Catalog NumberQuantity
LOV72225050 mg
Catalog number LOV722250
Price (MYR)
354.00
EA
Add to cart
Quantity:
50 mg
Price (MYR)
354.00
EA
Add to cart
Baricitinib is a highly potent JAK1 and JAK2 inhibitor (IC50 values are 5.9 and 5.7 nM, respectively). Also inhibits Tyk2 (IC50 = 53 nM). Displays >100-fold selectivity for JAK1/2 over JAK3 (IC50 = 53 nM for JAK3). Binds AAK1, BIKE, GAK and MPSK1 in vitro (Kd values are 17.2, 39.8, 134.4 and 68.5 nM, respectively). Inhibits IL-6-induced STAT phosphorylation and subsequent pro-inflammatory chemokine (MPC-1) and cytokine (IL-17 and IL-22) production in PBMCs and T-cells. Displays anti-inflammatory and disease modifying effects in the rat adjuvant arthritis model. In silico modelling predicts inhibition of SARS-CoV-2 cell entry.
Specifications
Chemical Name or Material1-(Ethylsulfonyl)-3-[4-(7 H-pyrrolo[2,3-d]pyrimidin-4-yl)-1 H-pyrazol-1-yl]-3-azetidineacetonitrile
CAS1187594-09-7
Recommended StorageStore at -20°C
Quantity50 mg
TargetJAK Kinase Inhibitors
Molecular FormulaC16H17N7O2S
Purity0.98
Unit SizeEA