For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Have Questions?
Catalog Number
Quantity
LOV511221BT
25 Tablets
Catalog number LOV511221BT
Price (MYR)
531.00
PK
Add to cart
Quantity:
25 Tablets
Price (MYR)
531.00
PK
Add to cart
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
DPD No. 4 Tablets
Available in packs of 250 and 500
New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
‘BT’ is added to the end of the code to identify the new style of packaging
Simple, time-saving and practical
There are no safety risks if the tablets are used in line with the instructions supplied
Safety data sheets are available for all reagents
Specifications
For Use With (Application)Photometry Tablet Reagents
For Use With (Equipment)Test kits, colorimeters and spectrophotometers