Check Order Status
Iodine Determination Flask
Iodine Determination Flask
Witeg™ Iodine Determination Flask is an Erlenmeyer-style flask with a ground glass joint, a modified stopper, and a funnel-shaped top.
Have Questions?
Catalog Number
Capacity (Metric)
LOV2438250
250 mL
Catalog number
LOV2438250
Price (MYR)
34,415.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
34,415.00
EA
Add to cart
Very good chemical resistance
High temperature resistance
Minimal thermal expansion
Stopper handle projects above the liquid seal trough to facilitate removal
Ground glass joint prevents vapors from escaping, improving the accuracy of your analysis
Funnel-shaped top to ensure material entry and minimize spillage
Specifications
Capacity (Metric)
250 mL
Material
Borosilicate Glass (3.3)
Includes
Hollow Stopper with Long Stem