MC500, MD600 Colorimeter Sample Cells
MC500, MD600 Colorimeter Sample Cells
Actual product may vary
MC500, MD600 Colorimeter Sample Cells
For use with MC500, MD600 colorimeters
Have Questions?
Catalog NumberDiameter (Metric)Includes
LOV19762024 mm12 Pk. Sample cells, lids
Catalog number LOV197620
Price (MYR)
870.00
PK
Add to cart
Diameter (Metric):
24 mm
Includes:
12 Pk. Sample cells, lids
Price (MYR)
870.00
PK
Add to cart
For use with MC500, MD600 colorimeters
  • Material: glass
  • Volume: 10mL
  • With lids
Specifications
Diameter (Metric)24 mm
TypeColorimeter Sample Cell
For Use With (Equipment)SP600 Spectrophotometer
Includes12 Pk. Sample cells, lids
MaterialGlass
Volume (Metric)10 mL
Unit SizePK