SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood
SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood
Actual product may vary
SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Have Questions?
Catalog NumberCut Style
LBC9500400No Cutouts
Catalog number LBC9500400
Price (MYR)
10,445.00
EA
Add to cart
Cut Style:
No Cutouts
Price (MYR)
10,445.00
EA
Add to cart
  • Corrosion-resistant epoxy resins molded
  • Front edge with large radius to aerodynamically direct airflow into hood
  • Pre-drilled holes for venting and plumbing pass-throughs provided
  • Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
  • Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Specifications
For Use With (Equipment)Protector XL Laboratory Hood
IncludesNo Cutouts
Product TypeSpillStopper Work Surface (4 ft.)
MaterialEpoxy Resin
For Use With (Application)Laboratory Fume Hood Work
Cut StyleNo Cutouts
Unit SizeEA