Clear Chemical Trap Canister with Stand
Clear Chemical Trap Canister with Stand
Actual product may vary
Clear Chemical Trap Canister with Stand
For use with FreeZone Freeze Dry Systems.
Have Questions?
Catalog number LBC7815302
Price (MYR)
4,723.00
EA
Add to cart
Price (MYR)
4,723.00
EA
Add to cart
  • Clear polypropylene canister allows visual inspection of trap contents
  • W x D x H: 17.8 x 17.8 x 33.0cm (7.0 x 7.0 x 13.0 in.)
  • Weight: 2.3kg (5.0 lb)
Specifications
TypeCanister
No. of Shelves0
ColorClear
MaterialPolypropylene
For Use With (Equipment)Freeze Dry Systems
IncludesStand
Depth (English)7.0 in.
Depth (Metric)17.8 cm
Width (English)7.0 in.
Width (Metric)17.8 cm
Height (English)13.0 in.
Height (Metric)33.0 cm
Weight (English)5.0 lb.
Weight (Metric)2.3 kg
Unit SizeEA