Secondary Trap Canister and Inserts for CentriVap™ System
Secondary Trap Canister and Inserts for CentriVap™ System
Actual product may vary
Secondary Trap Canister and Inserts for CentriVap™ System
Help to provide secondary protection to vacuum pumps in evaporation systems
Have Questions?
Catalog NumberTypeWeight (English)Weight (Metric)
LBC7815200Solvent Trap Insert4 lb.1.8 kg
Catalog number LBC7815200
Price (MYR)
1,124.00
EA
Add to cart
Type:
Solvent Trap Insert
Weight (English):
4 lb.
Weight (Metric):
1.8 kg
Price (MYR)
1,124.00
EA
Add to cart
Help to provide secondary protection to vacuum pumps in evaporation systems
  • Clear polypropylene canister with mounting bracket, 17 in. (43cm) PVC tubing, and two clamps
  • Canister accommodates trap inserts listed in the table
Specifications
MaterialPolypropylene
For Use With (Equipment)CentriVap™ system
TypeSolvent Trap Insert
Weight (English)4 lb.
Weight (Metric)1.8 kg
Unit SizeEA