FreeZone™ Stoppering or Bulk Tray Dryer Accessories
FreeZone™ Stoppering or Bulk Tray Dryer Accessories
Actual product may vary
FreeZone™ Stoppering or Bulk Tray Dryer Accessories
Accessories for FreeZone Stoppering and Bulk Tray Dryers.
Have Questions?
Catalog NumberLength (English)Weight (English)
LBC775610012 in.5 lb.
Catalog number LBC7756100
Price (MYR)
4,108.00
EA
Add to cart
Length (English):
12 in.
Weight (English):
5 lb.
Price (MYR)
4,108.00
EA
Add to cart
Accessories for FreeZone Stoppering and Bulk Tray Dryers.
Support Grid Stainless steel with removable plastic grid Provides support for stoppering ampules and other small containers Holds 144-12mm dia. containers Use with Labconco Stoppering Tray and Bulk Tray Dryers Tray with Slide-out Bottom Stainless steel with separate bottom that slides out to allow glassware containers direct contact with shelf Use with Labconco Stoppering Tray and Bulk Tray Dryers Shelf Spacers Stainless steel Place on one or two emply shelves to assist stoppering of partial loads of small vials or bottles
Specifications
TypeTray
No. of Shelves0
MaterialStainless Steel
For Use With (Equipment)Freezone™ stoppering tray dryer
IncludesSlide out bottom
Width (English)12 in.
Width (Metric)30.5 cm
Dimensions (L x W)12 x 12 in. (35.6 x 30.5cm)
Length (English)12 in.
Length (Metric)35.6 cm
Weight (English)5 lb.
Weight (Metric)2.3 kg
Unit SizeEA