Replacement Lid Gasket
Replacement Lid Gasket
Actual product may vary
Replacement Lid Gasket
Replacement lids for use with FreeZone 1, 2.5 and 4.5 Liter Freeze Dry Systems.
Have Questions?
Catalog number LBC7688500
Price (MYR)
-
Replacement lids for use with FreeZone 1, 2.5 & 4.5 Liter Freeze Dry Systems.
Replacement rubber gasket allows the chamber lid of a FreeZone Freeze Dryer to seal to the chamber during the lyophilization process.
Specifications
TypeLid Gasket
Capacity (Metric)1, 2.5, 4.5 L
For Use With (Equipment)Bulk drying chamber
Unit SizeEA